Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM2 SIRNA Assays ›
Gene Symbol:
ABLIM2
Gene Name:
actin binding LIM protein family member 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 7965310 - 8158832 on Build GRCh38
UniGene ID:
Hs.233404
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131323
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130083.1
NP_001123555.1
2
228
NM_001130084.1
NP_001123556.1
2
228
NM_001130085.1
NP_001123557.1
2
228
NM_001130086.1
NP_001123558.1
2
228
NM_001130087.1
NP_001123559.1
2
228
NM_001130088.1
NP_001123560.1
2
228
NM_032432.4
NP_115808.3
2
228
XM_005248014.4
XP_005248071.1
2
199
XM_005248017.4
XP_005248074.1
2
199
XM_005248018.4
XP_005248075.1
2
198
XM_005248019.4
XP_005248076.1
2
198
XM_005248020.4
XP_005248077.1
2
198
XM_005248021.4
XP_005248078.1
2
197
XM_005248022.4
XP_005248079.1
2
198
XM_005248023.4
XP_005248080.1
2
198
XM_005248024.4
XP_005248081.1
2
197
XM_005248027.4
XP_005248084.1
2
197
XM_005248028.4
XP_005248085.1
2
194
XM_005248029.4
XP_005248086.1
2
196
XM_005248030.4
XP_005248087.1
2
196
XM_005248031.4
XP_005248088.1
2
198
XM_006713923.3
XP_006713986.1
2
199
XM_006713924.3
XP_006713987.1
2
198
XM_006713925.3
XP_006713988.1
2
199
XM_006713927.3
XP_006713990.1
2
197
XM_011513586.2
XP_011511888.1
2
203
XM_017008706.1
XP_016864195.1
2
200
XM_017008707.1
XP_016864196.1
2
209
XM_017008708.1
XP_016864197.1
2
198
XM_017008709.1
XP_016864198.1
2
199
XM_017008710.1
XP_016864199.1
2
198
XM_017008711.1
XP_016864200.1
2
198
XM_017008712.1
XP_016864201.1
2
198
XM_017008713.1
XP_016864202.1
2
197
XM_017008714.1
XP_016864203.1
2
209
XM_017008715.1
XP_016864204.1
2
198
XM_017008716.1
XP_016864205.1
2
197
XM_017008717.1
XP_016864206.1
2
197
XM_017008718.1
XP_016864207.1
2
209
XM_017008719.1
XP_016864208.1
2
197
XM_017008720.1
XP_016864209.1
2
196
XM_017008721.1
XP_016864210.1
2
196
XM_017008722.1
XP_016864211.1
2
197
XM_017008723.1
XP_016864212.1
2
209
XM_017008724.1
XP_016864213.1
2
209
XM_017008725.1
XP_016864214.1
2
209
GenBank mRNA
AJ748600.1
-
2
150
AJ748601.1
-
2
150
AK094754.1
-
2
207
AK301060.1
-
2
191
BC067214.1
-
2
240
BC122567.1
-
2
179
40085570
DQ413176.1
-
2
105
DQ413177.1
-
1
85
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
actin cytoskeleton organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0