Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM2 SIRNA Assays ›
Gene Symbol:
ABLIM2
Gene Name:
actin binding LIM protein family member 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 7965310 - 8158832 on Build GRCh38
UniGene ID:
Hs.233404
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131325
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130083.1
NP_001123555.1
21
2796
NM_001130084.1
NP_001123556.1
20
2694
NM_001130085.1
NP_001123557.1
18
2577
NM_001130086.1
NP_001123558.1
18
2538
NM_001130087.1
NP_001123559.1
17
2454
NM_001286688.1
NP_001273617.1
15
2058
NM_032432.4
NP_115808.3
16
2424
XM_005248014.4
XP_005248071.1
23
2920
XM_005248017.4
XP_005248074.1
21
2803
XM_005248018.4
XP_005248075.1
22
2799
XM_005248019.4
XP_005248076.1
21
2766
XM_005248020.4
XP_005248077.1
21
2763
XM_005248021.4
XP_005248078.1
20
2681
XM_005248022.4
XP_005248079.1
20
2664
XM_005248023.4
XP_005248080.1
19
2646
XM_005248024.4
XP_005248081.1
18
2546
XM_005248027.4
XP_005248084.1
18
2507
XM_005248028.4
XP_005248085.1
17
2423
XM_005248029.4
XP_005248086.1
16
2392
XM_005248030.4
XP_005248087.1
16
2389
XM_006713923.3
XP_006713986.1
21
2800
XM_006713924.3
XP_006713987.1
22
2796
XM_006713925.3
XP_006713988.1
20
2680
XM_006713927.3
XP_006713990.1
17
2423
XM_011513586.2
XP_011511888.1
22
2914
XM_017008706.1
XP_016864195.1
23
2918
XM_017008707.1
XP_016864196.1
21
2813
XM_017008708.1
XP_016864197.1
20
2769
XM_017008709.1
XP_016864198.1
20
2701
XM_017008710.1
XP_016864199.1
21
2697
XM_017008711.1
XP_016864200.1
19
2670
XM_017008712.1
XP_016864201.1
19
2667
XM_017008713.1
XP_016864202.1
19
2648
XM_017008714.1
XP_016864203.1
19
2660
XM_017008715.1
XP_016864204.1
18
2592
XM_017008716.1
XP_016864205.1
19
2582
XM_017008717.1
XP_016864206.1
19
2579
XM_017008718.1
XP_016864207.1
19
2591
XM_017008719.1
XP_016864208.1
18
2549
XM_017008720.1
XP_016864209.1
19
2539
XM_017008721.1
XP_016864210.1
18
2509
XM_017008722.1
XP_016864211.1
17
2492
XM_017008724.1
XP_016864213.1
17
2438
XM_017008726.1
XP_016864215.1
17
2256
XM_017008727.1
XP_016864216.1
15
2139
GenBank mRNA
AB058711.1
-
17
2336
AK094754.1
-
17
2433
AK094798.1
-
15
2028
AK097003.1
-
2
998
AL834195.1
-
7
1791
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
actin cytoskeleton organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-wqf8l:80/100.66.76.116:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline