Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271287.1 | NP_001258216.1 | 4 | 470 | ||
NM_001271288.1 | NP_001258217.1 | 4 | 467 | |||
NM_001271318.1 | NP_001258247.1 | 6 | 630 | |||
NM_032434.3 | NP_115810.2 | 5 | 529 | |||
GenBank mRNA | AB058708.1 | - | 3 | 288 | ||
AK057028.1 | - | 5 | 439 | |||
AK294055.1 | - | 5 | 635 | |||
AK316011.1 | - | 6 | 490 | |||
AK316299.1 | - | 4 | 384 | |||
AL514772.3 | - | 5 | 446 | |||
BC043221.2 | - | 4 | 378 | 5295402 | ||
BC093041.1 | - | 4 | 397 | 6173398 | ||
BP251717.1 | - | 5 | 529 | |||
BX647996.1 | - | 4 | 376 |