Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143774.1 | NP_001137246.1 | 2 | 285 | ||
NM_032439.3 | NP_115815.2 | 2 | 505 | |||
XM_011540275.2 | XP_011538577.1 | 3 | 567 | |||
XM_011540276.2 | XP_011538578.1 | 2 | 1398 | |||
XM_017016782.1 | XP_016872271.1 | 3 | 386 | |||
XM_017016783.1 | XP_016872272.1 | 3 | 1489 | |||
GenBank mRNA | AK054956.1 | - | 2 | 309 | ||
AL365474.1 | - | 2 | 285 | |||
AL834339.1 | - | 2 | 506 | |||
AY358162.1 | - | 1 | 135 | |||
BC011268.2 | - | 2 | 344 | 4156912 | ||
CR749429.1 | - | 3 | 534 |