Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015877.1 | NP_001015877.1 | 4 | 512 | ||
NM_032335.3 | NP_115711.2 | 4 | 512 | |||
NM_032458.2 | NP_115834.1 | 4 | 512 | |||
GenBank mRNA | AB058726.1 | - | 4 | 398 | ||
AK290095.1 | - | 4 | 483 | |||
AK303369.1 | - | 3 | 375 | |||
AL832725.1 | - | 4 | 505 | |||
AY157622.1 | - | 4 | 512 | |||
BC005994.1 | - | 4 | 451 | 4064169 | ||
CD685336.1 | - | 4 | 520 | |||
DR000282.1 | - | 4 | 547 |