Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACRBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032489.2 | NP_115878.2 | 10 | 1663 | ||
GenBank mRNA | AB051833.1 | - | 10 | 1663 | ||
AF085884.1 | - | 2 | 283 | |||
AK302290.1 | - | 10 | 1561 | |||
AK311572.1 | - | 10 | 1470 | |||
AK312782.1 | - | 10 | 1681 | |||
BC033010.2 | - | 10 | 1660 | 4822765 |