Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080156.2 | NP_001073625.1 | 6 | 946 | ||
NM_001080157.1 | NP_001073626.1 | 7 | 1301 | |||
NM_001319850.1 | NP_001306779.1 | 11 | 1472 | |||
NM_001319851.1 | NP_001306780.1 | 5 | 1165 | |||
NM_001319852.1 | NP_001306781.1 | 6 | 946 | |||
NM_032496.3 | NP_115885.2 | 8 | 1227 | |||
XM_005269083.2 | XP_005269140.1 | 6 | 611 | |||
XM_005269084.2 | XP_005269141.1 | 5 | 544 | |||
XM_005269085.2 | XP_005269142.1 | 5 | 518 | |||
XM_011538656.2 | XP_011536958.1 | 10 | 1264 | |||
XM_011538657.2 | XP_011536959.1 | 7 | 1048 | |||
XM_011538658.2 | XP_011536960.1 | 7 | 1047 | |||
XM_011538659.2 | XP_011536961.1 | 10 | 1264 | |||
XM_017019800.1 | XP_016875289.1 | 7 | 1484 | |||
XM_017019801.1 | XP_016875290.1 | 6 | 611 | |||
XM_017019802.1 | XP_016875291.1 | 5 | 544 | |||
XM_017019803.1 | XP_016875292.1 | 6 | 611 | |||
XM_017019804.1 | XP_016875293.1 | 5 | 544 | |||
GenBank mRNA | AB030239.1 | - | 6 | 698 | ||
AB051853.1 | - | 8 | 1205 | |||
AK057573.1 | - | 5 | 818 | |||
AK092763.1 | - | 7 | 1292 | |||
AK300910.1 | - | 5 | 811 | |||
AK301095.1 | - | 6 | 598 | |||
AL713803.1 | - | 2 | 93 | |||
BC006107.1 | - | 11 | 1472 | 3010091 | ||
BC011820.1 | - | 6 | 672 |