Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184743.1 | NP_001171672.1 | 2, 3 | 844 | ||
NM_032507.3 | NP_115896.1 | 2, 3 | 824 | |||
XM_006715228.2 | XP_006715291.1 | 1, 2 | 501 | |||
XM_011514944.2 | XP_011513246.1 | 1, 2 | 501 | |||
XM_017011361.1 | XP_016866850.1 | 1, 2 | 501 | |||
XM_017011362.1 | XP_016866851.1 | 2, 3 | 617 | |||
XM_017011363.1 | XP_016866852.1 | 2, 3 | 800 | |||
GenBank mRNA | AK223446.1 | - | 2, 3 | 809 | ||
AK291228.1 | - | 2, 3 | 789 | |||
BC032507.1 | - | 2, 3 | 774 | |||
BC069033.1 | - | 2, 3 | 670 | |||
BC128585.1 | - | 2, 3 | 671 | 40128523 | ||
D88259.1 | - | 1, 2 | 404 |