Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032510.3 | NP_115899.1 | 2 | 266 | ||
GenBank mRNA | AB044556.1 | - | 2 | 100 | ||
AB178535.1 | - | 2 | 100 | |||
AB178536.1 | - | 2 | 100 | |||
BC060797.1 | - | 2 | 266 | 30348280 |