Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083113.1 | NP_001076582.1 | 7 | 1702 | ||
NM_001195653.1 | NP_001182582.1 | 7 | 1726 | |||
NM_001195654.1 | NP_001182583.1 | 7 | 1718 | |||
NM_032527.4 | NP_115916.3 | 7 | 1805 | |||
NM_181485.2 | NP_852150.2 | 7 | 1675 | |||
GenBank mRNA | AB058750.3 | - | 7 | 1679 | ||
AK027878.1 | - | 7 | 1743 | |||
AK074961.1 | - | 6 | 2629 | |||
AK307368.1 | - | 3 | 610 | |||
BC019338.1 | - | 7 | 1693 | 2906985 | ||
BC032612.1 | - | 7 | 1665 | 5555076 |
Gene Symbol: LIME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305654.1 | NP_001292583.1 | 1 | 239 | ||
GenBank mRNA | AY203957.1 | - | 1 | 239 |