Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIRREL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161707.1 | NP_001155179.1 | 12, 13 | 1917 | ||
NM_032531.3 | NP_115920.1 | 12, 13 | 1917 | |||
XM_011543026.2 | XP_011541328.1 | 12, 13 | 1661 | |||
XM_011543027.2 | XP_011541329.1 | 13, 14 | 2122 | |||
XM_011543028.2 | XP_011541330.1 | 13, 14 | 1981 | |||
XM_011543030.2 | XP_011541332.1 | 12, 13 | 1658 | |||
XM_011543031.2 | XP_011541333.1 | 13, 14 | 2116 | |||
XM_017018418.1 | XP_016873907.1 | 12, 13 | 2212 | |||
XM_017018420.1 | XP_016873909.1 | 13, 14 | 1981 | |||
GenBank mRNA | AB058770.1 | - | 12, 13 | 1806 | ||
AF480410.1 | - | 12, 13 | 1556 | |||
AY358743.1 | - | 12, 13 | 1857 | |||
AY358760.1 | - | 1, 2 | 93 | |||
BC101775.1 | - | 12, 13 | 1642 | 8069281 | ||
BC101801.1 | - | 12, 13 | 1642 | 8069307 |