Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144003.2 | NP_001137475.1 | 5 | 3711 | ||
NM_001144004.2 | NP_001137476.1 | 5 | 3699 | |||
NM_001144005.2 | NP_001137477.1 | 5 | 3721 | |||
NM_001144006.2 | NP_001137478.1 | 3 | 3931 | |||
NM_001144008.2 | NP_001137480.1 | 3 | 3943 | |||
NM_001144009.2 | NP_001137481.1 | 2 | 3784 | |||
NM_001144010.2 | NP_001137482.1 | 3 | 3921 | |||
NM_032539.4 | NP_115928.1 | 5 | 4194 | |||
XM_005262342.3 | XP_005262399.1 | 5 | 4796 | |||
XM_005262343.3 | XP_005262400.1 | 5 | 4806 | |||
XM_005262344.3 | XP_005262401.1 | 5 | 4784 | |||
XM_005262345.3 | XP_005262402.1 | 4 | 4647 | |||
GenBank mRNA | CR627412.1 | - | 2 | 5233 |