Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256267.1 | NP_001243196.1 | 3 | 315 | ||
NM_001256268.1 | NP_001243197.1 | 3 | 484 | |||
NM_032578.3 | NP_115967.2 | 2 | 420 | |||
NR_045663.3 | 2 | 420 | ||||
XM_017016833.1 | XP_016872322.1 | 3 | 756 | |||
XM_017016834.1 | XP_016872323.1 | 2 | 214 | |||
GenBank mRNA | AF328296.1 | - | 2 | 615 | ||
AK027343.1 | - | 2 | 358 | |||
AL832002.2 | - | 2 | 343 | |||
AL834247.2 | - | 2 | 342 | |||
BC142609.1 | - | 2 | 292 | |||
BC144334.1 | - | 2 | 182 | |||
BX647637.1 | - | 2 | 352 | |||
BX647640.1 | - | 3 | 309 |