Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256267.1 | NP_001243196.1 | 3 | 1042 | ||
NM_001256268.1 | NP_001243197.1 | 3 | 1211 | |||
NM_032578.3 | NP_115967.2 | 2 | 1147 | |||
NR_045663.3 | 2 | 1147 | ||||
XM_017016833.1 | XP_016872322.1 | 3 | 1483 | |||
XM_017016834.1 | XP_016872323.1 | 2 | 941 | |||
GenBank mRNA | AF328296.1 | - | 2 | 1342 | ||
AK027343.1 | - | 2 | 1085 | |||
AL832002.2 | - | 2 | 1070 | |||
AL834247.2 | - | 2 | 1069 | |||
BC142609.1 | - | 2 | 1019 | |||
BC144334.1 | - | 2 | 909 | |||
BX647637.1 | - | 2 | 1079 | |||
BX647640.1 | - | 3 | 1035 | |||
BX648564.1 | - | 4 | 1475 |