Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256267.1 | NP_001243196.1 | 8 | 1590 | ||
NM_001256268.1 | NP_001243197.1 | 11 | 1999 | |||
NM_032578.3 | NP_115967.2 | 7 | 1695 | |||
NR_045662.3 | 7 | 830 | ||||
NR_045663.3 | 7 | 1695 | ||||
XM_017016833.1 | XP_016872322.1 | 8 | 2031 | |||
XM_017016834.1 | XP_016872323.1 | 7 | 1489 | |||
GenBank mRNA | AF328296.1 | - | 7 | 1890 | ||
AK027343.1 | - | 7 | 1633 | |||
AL832002.2 | - | 7 | 1618 | |||
AL832379.1 | - | 7 | 792 | |||
AL834247.2 | - | 7 | 1617 | |||
BC142609.1 | - | 7 | 1567 | |||
BC144334.1 | - | 7 | 1457 | |||
BX647637.1 | - | 7 | 1627 | |||
BX647640.1 | - | 8 | 1583 | |||
BX648564.1 | - | 9 | 2023 |