Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032587.3 | NP_115976.2 | 3 | 2982 | ||
XM_017009989.1 | XP_016865478.1 | 2 | 2535 | |||
XM_017009990.1 | XP_016865479.1 | 4 | 3216 | |||
GenBank mRNA | AF356193.1 | - | 3 | 3106 | ||
BC093825.1 | - | 3 | 2929 | 7939670 |