Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005366.1 | NP_001005366.1 | 14 | 2353 | ||
NM_032590.4 | NP_115979.3 | 14 | 2143 | |||
XM_005253955.3 | XP_005254012.1 | 14 | 2353 | |||
XM_005253956.3 | XP_005254013.1 | 14 | 1950 | |||
XM_005253961.4 | XP_005254018.1 | 3 | 438 | |||
XM_011538867.2 | XP_011537169.1 | 14 | 3396 | |||
XM_011538868.2 | XP_011537170.1 | 14 | 3392 | |||
XM_011538869.2 | XP_011537171.1 | 14 | 3394 | |||
XM_011538875.2 | XP_011537177.1 | 3 | 438 | |||
GenBank mRNA | AB031230.1 | - | 4 | 602 | ||
AJ459424.1 | - | 14 | 2115 | |||
AK027440.1 | - | 4 | 532 | |||
AK027692.1 | - | 1 | 48 | |||
AK127328.1 | - | 14 | 2353 | |||
AK299827.1 | - | 3 | 448 | |||
AK316470.1 | - | 3 | 450 | |||
AL133572.1 | - | 10 | 1656 | |||
BC115380.1 | - | 14 | 2082 | 40029132 |