Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005366.1 | NP_001005366.1 | 23 | 4343 | ||
NM_032590.4 | NP_115979.3 | 23 | 4425 | |||
XM_005253955.3 | XP_005254012.1 | 23 | 4635 | |||
XM_005253956.3 | XP_005254013.1 | 23 | 4232 | |||
XM_005253961.4 | XP_005254018.1 | 12 | 2720 | |||
XM_011538868.2 | XP_011537170.1 | 23 | 5683 | |||
XM_011538869.2 | XP_011537171.1 | 24 | 5507 | |||
XM_011538875.2 | XP_011537177.1 | 12 | 2729 | |||
GenBank mRNA | AB031230.1 | - | 15 | 2906 | ||
AK027440.1 | - | 14 | 2701 | |||
AK027692.1 | - | 10 | 2330 | |||
AK074718.1 | - | 9 | 1800 | |||
AK127328.1 | - | 23 | 4343 | |||
AL133572.1 | - | 15 | 6220 | |||
BC008735.2 | - | 9 | 2195 | |||
BC115379.1 | - | 13 | 2415 | |||
BC115380.1 | - | 16 | 2655 | 40029132 |