Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126328.2 | NP_001119800.1 | 6 | 1291 | ||
NM_032622.2 | NP_116011.2 | 5 | 1022 | |||
XM_005265785.4 | XP_005265842.1 | 6 | 1449 | |||
XM_017008776.1 | XP_016864265.1 | 7 | 1510 | |||
GenBank mRNA | AF237782.1 | - | 5 | 953 | ||
AK056823.1 | - | 6 | 1264 | |||
AK294611.1 | - | 6 | 820 | |||
AY358326.1 | - | 5 | 933 | |||
BC022983.1 | - | 6 | 1167 | 4995278 | ||
BC034737.1 | - | 5 | 942 | 5194423 |