Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126328.2 | NP_001119800.1 | 10 | 2313 | ||
NM_032622.2 | NP_116011.2 | 9 | 2044 | |||
XM_005265785.4 | XP_005265842.1 | 10 | 2471 | |||
XM_017008776.1 | XP_016864265.1 | 11 | 2532 | |||
GenBank mRNA | AF237782.1 | - | 9 | 1975 | ||
AK056823.1 | - | 10 | 2286 | |||
AK294611.1 | - | 10 | 1842 | |||
AY358326.1 | - | 9 | 1955 | |||
BC022983.1 | - | 10 | 2189 | 4995278 | ||
BC034737.1 | - | 9 | 1964 | 5194423 | ||
BC094772.1 | - | 6 | 1492 |