Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201484.1 | NP_001188413.1 | 13 | 2618 | ||
NM_032634.3 | NP_116023.2 | 11 | 3771 | |||
NM_152850.3 | NP_690577.2 | 12 | 2166 | |||
XM_005251619.3 | XP_005251676.1 | 11 | 3396 | |||
XM_017015222.1 | XP_016870711.1 | 12 | 3888 | |||
XM_017015223.1 | XP_016870712.1 | 12 | 2547 | |||
XM_017015224.1 | XP_016870713.1 | 12 | 2811 | |||
GenBank mRNA | AK074064.1 | - | 5 | 5154 | ||
AK090433.1 | - | 4 | 6358 | |||
AL833956.1 | - | 11 | 3804 | |||
AY358472.1 | - | 10 | 3377 | |||
BC001030.2 | - | 9 | 1496 | |||
BC013987.2 | - | 10 | 1582 | |||
BC029271.1 | - | 10 | 3042 | |||
BC036916.1 | - | 11 | 2131 | 5203414 | ||
BC065282.1 | - | 13 | 2289 | 6172460 |