Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYROXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032709.2 | NP_116098.2 | 1 | 158 | ||
XM_011540293.2 | XP_011538595.1 | 1 | 17 | |||
XM_017016835.1 | XP_016872324.1 | 1 | 119 | |||
XM_017016836.1 | XP_016872325.1 | 1 | 119 | |||
XM_017016837.1 | XP_016872326.1 | 1 | 119 | |||
GenBank mRNA | AK074429.1 | - | 1 | 119 | ||
AK075265.1 | - | 1 | 96 | |||
AK225637.1 | - | 1 | 112 | |||
AK295699.1 | - | 1 | 131 | |||
BC006131.1 | - | 1 | 121 | 3626506 |