Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYROXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032709.2 | NP_116098.2 | 16 | 1923 | ||
XM_011540293.2 | XP_011538595.1 | 17 | 3345 | |||
XM_011540301.2 | XP_011538603.1 | 9 | 2759 | |||
XM_017016838.1 | XP_016872327.1 | 16 | 4119 | |||
XM_017016839.1 | XP_016872328.1 | 14 | 1976 | |||
XM_017016840.1 | XP_016872329.1 | 14 | 1830 | |||
XM_017016841.1 | XP_016872330.1 | 13 | 2137 | |||
GenBank mRNA | AF461898.1 | - | 1 | 2395 | ||
AK074429.1 | - | 16 | 1884 | |||
AK075265.1 | - | 16 | 1861 | |||
AK123713.1 | - | 6 | 1880 | |||
AK225637.1 | - | 16 | 1877 | |||
BC006131.1 | - | 16 | 1886 | 3626506 |