Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032726.3 | NP_116115.1 | 6 | 947 | ||
XM_005246913.2 | XP_005246970.1 | 6 | 909 | |||
XM_011512012.1 | XP_011510314.1 | 6 | 909 | |||
XM_017005114.1 | XP_016860603.1 | 6 | 850 | |||
XM_017005115.1 | XP_016860604.1 | 6 | 909 | |||
XM_017005116.1 | XP_016860605.1 | 6 | 909 | |||
XM_017005117.1 | XP_016860606.1 | 6 | 909 | |||
XM_017005118.1 | XP_016860607.1 | 6 | 1736 | |||
XM_017005119.1 | XP_016860608.1 | 3 | 207 | |||
GenBank mRNA | AB209599.1 | - | 1 | 353 | ||
AK023083.1 | - | 5 | 643 | |||
AK057186.1 | - | 6 | 813 | |||
AK223203.1 | - | 6 | 813 | |||
AK299458.1 | - | 6 | 814 | |||
AY512961.1 | - | 6 | 857 | |||
BC006355.2 | - | 6 | 803 | 4124286 |