Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032726.3 | NP_116115.1 | 16 | 3133 | ||
XM_005246913.2 | XP_005246970.1 | 15 | 3173 | |||
XM_011512012.1 | XP_011510314.1 | 16 | 3269 | |||
XM_017005114.1 | XP_016860603.1 | 16 | 3210 | |||
XM_017005115.1 | XP_016860604.1 | 17 | 2929 | |||
XM_017005116.1 | XP_016860605.1 | 17 | 3191 | |||
XM_017005117.1 | XP_016860606.1 | 15 | 3116 | |||
XM_017005118.1 | XP_016860607.1 | 16 | 4096 | |||
XM_017005119.1 | XP_016860608.1 | 13 | 2567 | |||
XM_017005120.1 | XP_016860609.1 | 9 | 1979 | |||
GenBank mRNA | AB209599.1 | - | 8 | 4249 | ||
AK023083.1 | - | 14 | 3172 | |||
AK223203.1 | - | 16 | 2999 | |||
BC006355.2 | - | 16 | 2989 | 4124286 |