Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032737.3 | NP_116126.3 | 12 | 3988 | ||
GenBank mRNA | AK129641.1 | - | 2 | 923 | ||
M94362.1 | - | 11 | 3630 |