Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZYX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010972.1 | NP_001010972.1 | 4 | 583 | ||
NM_003461.4 | NP_003452.1 | 4 | 586 | |||
XM_017012587.1 | XP_016868076.1 | 4 | 676 | |||
GenBank mRNA | AK302309.1 | - | 3 | 394 | ||
AK316227.1 | - | 3 | 278 | |||
BC002323.2 | - | 4 | 458 | |||
BC008743.2 | - | 4 | 550 | 3345843 | ||
BC009360.2 | - | 4 | 495 | 4301542 | ||
BC010031.2 | - | 4 | 483 | 3506023 | ||
BC017183.2 | - | 3 | 112 | |||
BU844538.1 | - | 552 | ||||
CR457431.1 | - | 3 | 428 | |||
U15158.1 | - | 2 | 191 | |||
X94991.1 | - | 3 | 438 | |||
X95735.1 | - | 4 | 499 |