Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032765.2 | NP_116154.1 | 1 | 709 | ||
XM_005266000.3 | XP_005266057.1 | 1 | 751 | |||
XM_017009991.1 | XP_016865480.1 | 1 | 751 | |||
XM_017009992.1 | XP_016865481.1 | 1 | 751 | |||
XM_017009993.1 | XP_016865482.1 | 1 | 751 | |||
XM_017009994.1 | XP_016865483.1 | 1 | 751 | |||
GenBank mRNA | AB209243.1 | - | 1 | 727 | ||
AK054802.1 | - | 1 | 686 | |||
BC007372.2 | - | 1 | 709 | 3636274 | ||
JX896146.1 | - | 1 | 404 |