Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032765.2 | NP_116154.1 | 2 | 1192 | ||
GenBank mRNA | AB209243.1 | - | 1 | 3715 | ||
AK054802.1 | - | 2 | 1169 | |||
BC007372.2 | - | 2 | 1192 | 3636274 | ||
BC034154.1 | - | 1 | 1147 |