Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258452.1 | NP_001245381.1 | 2 | 192 | ||
NM_001258453.1 | NP_001245382.1 | 3 | 264 | |||
NM_032807.4 | NP_116196.3 | 3 | 280 | |||
NM_178150.2 | NP_835363.1 | 2 | 166 | |||
XM_011519748.2 | XP_011518050.1 | 4 | 860 | |||
XM_017016842.1 | XP_016872331.1 | 3 | 1407 | |||
XM_017016843.1 | XP_016872332.1 | 4 | 791 | |||
XM_017016844.1 | XP_016872333.1 | 2 | 176 | |||
XM_017016846.1 | XP_016872335.1 | 1 | 22 | |||
GenBank mRNA | AF380349.1 | - | 3 | 280 | ||
AF454502.1 | - | 2 | 110 | |||
AF456237.1 | - | 2 | 122 | |||
AK095343.1 | - | 2 | 140 | |||
AK122753.1 | - | 2 | 192 | |||
AK226128.1 | - | 2 | 690 | |||
BC110884.1 | - | 3 | 189 | 6147373 | ||
BC113375.1 | - | 2 | 113 | 8322427 | ||
BC113377.1 | - | 2 | 113 | 8322429 | ||
DA326341.1 | - | 2 | 179 |