Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318391.1 | NP_001305320.1 | 2 | 128 | ||
NM_032830.2 | NP_116219.2 | 2 | 182 | |||
GenBank mRNA | AB075868.2 | - | 2 | 120 | ||
AK027634.1 | - | 2 | 134 | |||
AK027675.1 | - | 2 | 131 | |||
AK074795.1 | - | 2 | 148 | |||
AK310104.1 | - | 2 | 75 | |||
BC009348.2 | - | 2 | 114 | 4131625 | ||
BX647265.1 | - | 2 | 96 | |||
DA277545.1 | - | 2 | 128 | |||
DC368305.1 | - | 2 | 182 |