Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318391.1 | NP_001305320.1 | 5 | 534 | ||
NM_032830.2 | NP_116219.2 | 5 | 588 | |||
GenBank mRNA | AB075868.2 | - | 5 | 526 | ||
AK027419.1 | - | 2 | 143 | |||
AK027445.1 | - | 3 | 211 | |||
AK027634.1 | - | 5 | 540 | |||
AK027675.1 | - | 5 | 537 | |||
AK074795.1 | - | 5 | 554 | |||
AK310104.1 | - | 5 | 481 | |||
BC000167.2 | - | 1 | 33 | |||
BC009348.2 | - | 5 | 520 | 4131625 | ||
BX647265.1 | - | 5 | 502 |