Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077268.1 | NP_001070736.1 | 3, 4 | 958 | ||
NM_001258420.1 | NP_001245349.1 | 3, 4 | 958 | |||
NM_001258421.1 | NP_001245350.1 | 3, 4 | 761 | |||
NM_032850.4 | NP_116239.3 | 4, 5 | 627 | |||
XM_017022683.1 | XP_016878172.1 | 2, 3 | 1603 | |||
XM_017022684.1 | XP_016878173.1 | 3, 4 | 945 | |||
GenBank mRNA | AB209224.1 | - | 2360 | |||
AF445414.1 | - | 4, 5 | 469 | |||
AK027746.1 | - | 3, 4 | 390 | |||
AK122779.1 | - | 3, 4 | 748 | |||
AK225553.1 | - | 3, 4 | 381 | |||
BC021092.1 | - | 3, 4 | 255 | 4662224 | ||
DB012963.1 | - | 3, 4 | 376 |