Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193308.1 | NP_001180237.1 | 10 | 1125 | ||
NM_032872.2 | NP_116261.1 | 10 | 1089 | |||
XM_005246022.3 | XP_005246079.1 | 10 | 1280 | |||
XM_006710990.1 | XP_006711053.1 | 10 | 1244 | |||
XM_006710991.1 | XP_006711054.1 | 9 | 879 | |||
GenBank mRNA | AK027902.1 | - | 10 | 1059 | ||
AK074154.1 | - | 6 | 1718 | |||
AK096437.1 | - | 8 | 1686 | |||
AK097172.1 | - | 5 | 1229 | |||
AK293038.1 | - | 9 | 1391 | |||
AY037157.1 | - | 10 | 1052 | |||
BC009224.2 | - | 8 | 656 | |||
BC015764.1 | - | 9 | 868 | 4851468 | ||
BC035725.1 | - | 10 | 1032 | 5722683 |