Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032890.3 | NP_116279.2 | 10 | 2098 | ||
XM_005273335.2 | XP_005273392.1 | 8 | 1939 | |||
XM_006711592.2 | XP_006711655.1 | 9 | 2886 | |||
XM_006711594.3 | XP_006711657.1 | 8 | 1594 | |||
XM_011510072.2 | XP_011508374.1 | 9 | 1938 | |||
XM_011510073.2 | XP_011508375.1 | 9 | 2416 | |||
XM_011510074.2 | XP_011508376.1 | 9 | 1957 | |||
XM_011510075.2 | XP_011508377.1 | 9 | 2444 | |||
XM_011510077.2 | XP_011508379.1 | 8 | 1880 | |||
XM_017002611.1 | XP_016858100.1 | 8 | 2354 | |||
XM_017002612.1 | XP_016858101.1 | 9 | 1763 | |||
XM_017002613.1 | XP_016858102.1 | 7 | 1723 | |||
GenBank mRNA | AK056569.1 | - | 3 | 902 | ||
BC011542.1 | - | 1 | 233 |