Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136049.2 | NP_001129521.2 | 15 | 1702 | ||
NM_033029.3 | NP_149018.2 | 14 | 1591 | |||
NR_026786.1 | 15 | 1581 | ||||
NR_026787.1 | 14 | 1470 | ||||
XM_011513270.1 | XP_011511572.1 | 15 | 1670 | |||
XM_011513271.1 | XP_011511573.1 | 15 | 1685 | |||
XM_017007428.1 | XP_016862917.1 | 14 | 1559 | |||
GenBank mRNA | AJ312398.1 | - | 15 | 1581 | ||
AJ312399.1 | - | 14 | 1591 | |||
AK299119.1 | - | 15 | 1669 | |||
AM920777.1 | - | 15 | 1680 | |||
AM920778.1 | - | 14 | 1413 | |||
DB054915.1 | - | 3 | 265 |