Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003302.2 | NP_003293.2 | 8 | 1417 | ||
GenBank mRNA | AB628085.1 | - | 8 | 1184 | ||
AB628086.1 | - | 8 | 1197 | |||
AB628087.1 | - | 7 | 1119 | |||
AB628088.1 | - | 8 | 1000 | |||
AF000974.1 | - | 8 | 1406 | |||
AF025437.1 | - | 5 | 590 | |||
AJ001902.1 | - | 8 | 1319 | |||
AK056773.1 | - | 7 | 1421 | |||
AK291906.1 | - | 8 | 1354 | |||
BC002680.2 | - | 8 | 1310 | 3610413 | ||
BC004249.1 | - | 8 | 1322 | 3621877 | ||
BC004999.1 | - | 8 | 1316 | 2958142 | ||
BC021540.1 | - | 8 | 1322 | 3622017 | ||
BC028985.1 | - | 8 | 1329 | 5122169 | ||
CR749505.1 | - | 7 | 2044 | |||
L40374.1 | - | 3 | 320 |