Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCAY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033064.4 | NP_149053.1 | 13 | 1750 | ||
GenBank mRNA | AB058775.1 | - | 13 | 1694 | ||
AK027889.1 | - | 13 | 1684 | |||
AK092309.1 | - | 13 | 1549 | |||
AK125457.1 | - | 13 | 1271 | |||
AK299522.1 | - | 14 | 1755 | |||
BC026217.1 | - | 13 | 1692 | 4153341 |