Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033103.4 | NP_149094.3 | 15 | 2948 | ||
GenBank mRNA | AF423421.1 | - | 15 | 2882 | ||
AK026894.1 | - | 1 | 710 | |||
AK095001.1 | - | 6 | 1718 | |||
AK098246.1 | - | 15 | 2918 | |||
AK126506.1 | - | 12 | 3216 | |||
AL831950.1 | - | 14 | 2796 | |||
BC036447.2 | - | 15 | 2918 | 4830913 |