Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033118.3 | NP_149109.1 | 3 | 315 | ||
GenBank mRNA | AF325549.1 | - | 2 | 213 | ||
AJ272502.1 | - | 3 | 321 | |||
BC069627.1 | - | 2 | 213 | 7262446 | ||
BC092413.1 | - | 3 | 315 | 30530456 | ||
BC127622.1 | - | 2 | 213 | 40085085 | ||
BX647635.1 | - | 3 | 298 |