Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033118.3 | NP_149109.1 | 13 | 2756 | ||
GenBank mRNA | AJ272502.1 | - | 13 | 2762 | ||
BC007753.1 | - | 6 | 1592 | |||
BC092413.1 | - | 13 | 2756 | 30530456 | ||
BX647635.1 | - | 13 | 4892 | |||
F22840.1 | - | 1 | 351 |