Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271082.1 | NP_001258011.1 | 11 | 1147 | ||
NM_033120.3 | NP_149111.1 | 10 | 968 | |||
XM_005248382.3 | XP_005248439.1 | 10 | 1232 | |||
XM_017010000.1 | XP_016865489.1 | 5 | 1418 | |||
GenBank mRNA | AB062887.1 | - | 10 | 877 | ||
AF358137.1 | - | 10 | 889 | |||
BC004940.1 | - | 5 | 496 | |||
BC012176.1 | - | 11 | 1147 | 4649935 |