Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033123.3 | NP_149114.2 | 4 | 581 | ||
NR_073075.1 | 4 | 584 | ||||
XM_005253519.2 | XP_005253576.1 | 4 | 574 | |||
XM_017020176.1 | XP_016875665.1 | 4 | 574 | |||
XM_017020177.1 | XP_016875666.1 | 4 | 574 | |||
XM_017020178.1 | XP_016875667.1 | 4 | 574 | |||
XM_017020179.1 | XP_016875668.1 | 4 | 569 | |||
XM_017020180.1 | XP_016875669.1 | 4 | 463 | |||
XM_017020181.1 | XP_016875670.1 | 4 | 528 | |||
GenBank mRNA | AF532185.1 | - | 4 | 506 | ||
AK292279.1 | - | 4 | 579 | |||
AK314516.1 | - | 4 | 537 | |||
AY035866.1 | - | 4 | 543 |