Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002006.2 | NP_001002006.1 | 6 | 1086 | ||
NM_001199086.1 | NP_001186015.1 | 6 | 1035 | |||
NM_001199087.1 | NP_001186016.1 | 6 | 1137 | |||
NM_001199088.1 | NP_001186017.1 | 6 | 1092 | |||
NM_033253.3 | NP_150278.2 | 5 | 906 | |||
GenBank mRNA | AF352326.1 | - | 1 | 44 | ||
AF356185.1 | - | 5 | 840 | |||
AF417165.1 | - | 6 | 1052 | |||
AK093234.1 | - | 2 | 189 | |||
AK093464.1 | - | 5 | 831 | |||
AK097422.1 | - | 5 | 895 | |||
AK097848.1 | - | 4 | 1300 | |||
AK302084.1 | - | 5 | 852 | |||
AK302200.1 | - | 6 | 981 | |||
AK302795.1 | - | 6 | 1121 | |||
BC136387.1 | - | 5 | 859 | 9020374 | ||
BC171761.1 | - | 5 | 859 | 9054414 |
Gene Symbol: NT5C1B-RDH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199103.1 | NP_001186032.1 | 5 | 912 | ||
NM_001199104.1 | NP_001186033.1 | 6 | 1086 | |||
GenBank mRNA | AK057444.1 | - | 6 | 1068 | ||
AK307653.1 | - | 5 | 896 |