Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272012.1 | NP_001258941.1 | 2 | 2370 | ||
NM_001272013.1 | NP_001258942.1 | 2 | 2490 | |||
NM_033397.3 | NP_203755.1 | 3 | 2682 | |||
XM_005270257.2 | XP_005270314.1 | 1 | 2246 | |||
GenBank mRNA | AB051541.1 | - | 2 | 2401 | ||
AK055608.1 | - | 1 | 1093 | |||
AK056526.1 | - | 2 | 2302 | |||
AK094802.1 | - | 1 | 1722 | |||
AY189283.1 | - | 1 | 586 | |||
AY358776.1 | - | 1 | 555 | |||
BC040536.1 | - | 2 | 2342 | 5265379 | ||
BC070108.1 | - | 3 | 2682 | 4837204 |