Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042632.2 | NP_001036097.1 | 4 | 2137 | ||
NM_001042633.2 | NP_001036098.1 | 5 | 2155 | |||
NM_033421.3 | NP_219489.1 | 4 | 2144 | |||
NM_152897.2 | NP_690857.1 | 5 | 892 | |||
XM_005260608.4 | XP_005260665.1 | 2 | 2219 | |||
XM_011529095.2 | XP_011527397.1 | 2 | 2123 | |||
GenBank mRNA | AF258570.1 | - | 1 | 1805 | ||
AK091359.1 | - | 3 | 2159 | |||
AK095851.1 | - | 3 | 2015 | |||
BC019823.1 | - | 5 | 865 | 4453502 |