Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTTNBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033427.2 | NP_219499.1 | 3 | 304 | ||
XM_005250634.4 | XP_005250691.1 | 3 | 439 | |||
XM_005250635.3 | XP_005250692.1 | 3 | 236 | |||
XM_011516615.2 | XP_011514917.1 | 3 | 2607 | |||
XM_011516616.2 | XP_011514918.1 | 3 | 236 | |||
XM_011516618.2 | XP_011514920.1 | 3 | 236 | |||
XM_017012706.1 | XP_016868195.1 | 3 | 236 | |||
XM_017012707.1 | XP_016868196.1 | 3 | 236 | |||
GenBank mRNA | AB051545.1 | - | 3 | 210 | ||
AF377960.1 | - | 3 | 304 | |||
AK091548.1 | - | 3 | 238 | |||
AK308081.1 | - | 3 | 236 | |||
BC106000.1 | - | 3 | 271 | 30528261 |