Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314044.1 | NP_001300973.1 | 4, 5 | 416 | ||
NM_001314045.1 | NP_001300974.1 | 4, 5 | 424 | |||
NM_001314048.1 | NP_001300977.1 | 3, 4 | 303 | |||
NM_033439.3 | NP_254274.1 | 4, 5 | 429 | |||
XM_011518061.2 | XP_011516363.1 | 3, 4 | 364 | |||
GenBank mRNA | AB024518.1 | - | 3, 4 | 368 | ||
AK313414.1 | - | 4, 5 | 408 | |||
AY905581.1 | - | 3, 4 | 351 | |||
BC047085.1 | - | 4, 5 | 406 | 5314040 | ||
BQ878541.1 | - | 3, 4 | 390 | |||
CR407619.1 | - | 3, 4 | 351 | |||
DA439244.1 | - | 4, 5 | 424 | |||
DA713547.1 | - | 4, 5 | 416 | |||
DB477860.1 | - | 4, 5 | 429 | |||
HQ641439.1 | - | 2, 3 | 240 |