Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199640.1 | NP_001186569.1 | 7 | 758 | ||
NM_001199641.1 | NP_001186570.1 | 5 | 506 | |||
NM_001314044.1 | NP_001300973.1 | 8 | 871 | |||
NM_001314045.1 | NP_001300974.1 | 8 | 879 | |||
NM_001314046.1 | NP_001300975.1 | 8 | 866 | |||
NM_001314047.1 | NP_001300976.1 | 8 | 853 | |||
NM_001314048.1 | NP_001300977.1 | 7 | 758 | |||
NM_033439.3 | NP_254274.1 | 8 | 884 | |||
XM_011518061.2 | XP_011516363.1 | 7 | 819 | |||
XM_017015285.1 | XP_016870774.1 | 10 | 1612 | |||
GenBank mRNA | AB024518.1 | - | 7 | 823 | ||
AK295908.1 | - | 5 | 487 | |||
AK303943.1 | - | 7 | 735 | |||
AL080211.1 | - | 1 | 184 | |||
BC047085.1 | - | 8 | 861 | 5314040 | ||
HQ641439.1 | - | 6 | 695 |