Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003940.1 | NP_001003940.1 | 5 | 1895 | ||
NM_001003942.1 | NP_001003942.1 | 3 | 1597 | |||
NM_001003943.2 | NP_001003943.1 | 4 | 1744 | |||
NM_033503.3 | NP_277038.1 | 4 | 1746 | |||
GenBank mRNA | AK024472.1 | - | 4 | 1756 | ||
BC060783.1 | - | 5 | 1893 | |||
BC070043.1 | - | 5 | 1893 | 30344794 |